Order Entry
Ireland
ContactUsLinkComponent
 
Analytical standard, Valifenalate, reference material, Honeywell Fluka™
  HONCR1111-25MG
 :  
Analytical standard, Valifenalate, reference material, Honeywell Fluka™
  HONCR1111-25MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    25 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    283159-90-0
  • Description:
    Valifenalate
  • Melting point:
    147 °C
  • Formula:
    C₁₉H₂₇ClN₂O₅
  • Storage temperature:
    Ambient
  • Description:
    Valifenalate
  • Boiling point:
    367 °C

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results