Order Entry
Ireland
ContactUsLinkComponent
 
Analytical standard, Fenthion oxon sulfone, reference material, Honeywell Fluka™
Catalog # HONCR1099-25MG
CAS Number:  
Analytical standard, Fenthion oxon sulfone, reference material, Honeywell Fluka™
Catalog # HONCR1099-25MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    25 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    14086-35-2
  • Description:
    Fenthion oxon sulfone
  • Melting point:
    78…79 °C
  • Formula:
    C₁₀H₁₅O₆PS
  • Storage temperature:
    2…8 °C
  • Description:
    Fenthion oxon sulfone

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results