Order Entry
Ireland
ContactUsLinkComponent
Analytical standard, Baquiloprim-d₆, reference material, Honeywell Fluka™
Analytical standard, Baquiloprim-d₆, reference material, Honeywell Fluka™
  HONCR1028-10MG
 :  
Analytical standard, Baquiloprim-d₆, reference material, Honeywell Fluka™
  HONCR1028-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1228182-50-0
  • Description:
    Baquiloprim-d₆
  • Melting point:
    233 °C
  • Formula:
    C₁₇D₆H₁₄N₆
  • Storage temperature:
    Ambient
  • Description:
    Baquiloprim-d₆

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results