Order Entry
Ireland
ContactUsLinkComponent
 
Nylon webbing sling, Protecta®, DBI-SALA®, 3M™
  CPSDAM450/100
undefined
Nylon webbing sling, Protecta®, DBI-SALA®, 3M™
  CPSDAM450/100

 

  • Description:
    Nylon webbing sling, 25×600 mm
  • Seq:
    01
  • Environmentally Preferable:
  • Weight:
    0,06 kg
  • Pk:
    1

 

 

Polyester webbing slings are made from top quality materials guaranteeing high strength and durability. Can be used as versatile temporary anchorage solution.

  • Durable and versatile
  • Four safety seams
  • Visible colour easy to see on site
 : EN 795, Class B