Order Entry
Ireland
ContactUsLinkComponent
Hydroxy-2-naphthalenylmethylphosphonic acid ≥95%, crystalline solid
Hydroxy-2-naphthalenylmethylphosphonic acid ≥95%, crystalline solid
Supplier:  Cayman Chemical
Certificates
Hydroxy-2-naphthalenylmethylphosphonic acid ≥95%, crystalline solid
Supplier:  Cayman Chemical

Specification Test Results

  • Purity:
    ≥95%
  • λmax:
    228 nm

Specification Test Results

Frequently Bought Together