Order Entry
Ireland
ContactUsLinkComponent
Nitrile gloves, Showa 6110PF
Nitrile gloves, Showa 6110PF
Supplier:  Showa
   Sustainable Options Available
Certificates
undefined
Nitrile gloves, Showa 6110PF
Supplier:  Showa
   Sustainable Options Available
Specifications
  • Catalog No:
  • 112-5637
  • BEST6110PFXS
  • 112-5638
  • 112-5640
  • 112-5641
  • 112-5639
  • Model:
  • Showa 6110PF
  • Showa 6110PF
  • Showa 6110PF
  • Showa 6110PF
  • Showa 6110PF
  • Showa 6110PF
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Environmentally Preferable:
  • Sustainable Materials, Safer Human and Environmental Health, Sustainable Packaging
  • Sustainable Materials, Safer Human and Environmental Health, Sustainable Packaging
  • Sustainable Materials, Safer Human and Environmental Health, Sustainable Packaging
  • Sustainable Materials, Safer Human and Environmental Health, Sustainable Packaging
  • Sustainable Materials, Safer Human and Environmental Health, Sustainable Packaging
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Smooth
  • Smooth
  • Smooth
  • Smooth
  • Smooth
  • Smooth
  • Cuff style:
  • Rolled
  • Rolled
  • Rolled
  • Rolled
  • Rolled
  • Rolled
  • EN norm:
  • EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
  • EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
  • EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
  • EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
  • EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
  • EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
  • Food contact approved:
  • +
  • +
  • +
  • +
  • +
  • +
  • Environmental certification:
  • Biodegradation test results acc. to the norm ASTM D5526. Green circle
  • Biodegradation test results acc. to the norm ASTM D5526. Green circle
  • Biodegradation test results acc. to the norm ASTM D5526. Green circle
  • Biodegradation test results acc. to the norm ASTM D5526. Green circle
  • Biodegradation test results acc. to the norm ASTM D5526. Green circle