Order Entry
Ireland
ContactUsLinkComponent
 
Anti-Tnfrsf16 Mouse Monoclonal Antibody [clone: MC192] Saporin
 :  Biosensis
 
undefined
Anti-Tnfrsf16 Mouse Monoclonal Antibody [clone: MC192] Saporin
 :  Biosensis

 

  • Catalog No:
  • BSENM-1755-50-IT
  • BSENM-1755-100-IT
  • BSENM-1755-25-IT
  • Antigen symbol:
  • Tnfrsf16
  • Tnfrsf16
  • Tnfrsf16
  • Antigen name:
  • NGFR/p75 neurotrophin receptor (p75NTR)
  • NGFR/p75 neurotrophin receptor (p75NTR)
  • NGFR/p75 neurotrophin receptor (p75NTR)
  • Antigen synonyms:
  • Low affinity neurotrophin receptor p75NTR|CD271|NGF receptor|p75 ICD|Gp80-LNGFR|Low-affinity nerve growth factor receptor
  • Low affinity neurotrophin receptor p75NTR|CD271|NGF receptor|p75 ICD|Gp80-LNGFR|Low-affinity nerve growth factor receptor
  • Low affinity neurotrophin receptor p75NTR|CD271|NGF receptor|p75 ICD|Gp80-LNGFR|Low-affinity nerve growth factor receptor
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Saporin
  • Saporin
  • Saporin
  • Clone:
  • MC192
  • MC192
  • MC192
  • Reactivity:
  • Rat
  • Rat
  • Rat
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • UniprotID #P07174, P20656
  • UniprotID #P07174, P20656
  • UniprotID #P07174, P20656
  • Isotype:
  • IgG1
  • IgG1
  • IgG1
  • Epitope:
  • Extracellular
  • Extracellular
  • Extracellular
  • Immunogen:
  • Rat NGF receptor (p75NTR)
  • Rat NGF receptor (p75NTR)
  • Rat NGF receptor (p75NTR)
  • Amino acid number:
  • 299 amino acids
  • 299 amino acids
  • 299 amino acids
  • Form:
  • Lyophillised
  • Lyophillised
  • Lyophillised
  • Molecular weight:
  • 30 kDa
  • 30 kDa
  • 30 kDa
  • Purification:
  • Conjugate was purified by ion-exchange chromatography. Purity >90% by non-reducing SDS-PAGE
  • Conjugate was purified by ion-exchange chromatography. Purity >90% by non-reducing SDS-PAGE
  • Conjugate was purified by ion-exchange chromatography. Purity >90% by non-reducing SDS-PAGE
  • Concentration:
  • 1 mg/ml
  • 1 mg/ml
  • 1 mg/ml
  • Storage buffer:
  • Reconstitute in sterile-filtered, ultrapure water to achieve a conjugate concentration of 1 mg/ml. Centrifuge to remove any insoluble material. Final buffer contains no preservatives.
  • Reconstitute in sterile-filtered, ultrapure water to achieve a conjugate concentration of 1 mg/ml. Centrifuge to remove any insoluble material. Final buffer contains no preservatives.
  • Reconstitute in sterile-filtered, ultrapure water to achieve a conjugate concentration of 1 mg/ml. Centrifuge to remove any insoluble material. Final buffer contains no preservatives.
  • Storage temperature:
  • 2…8 °C (lyophilised);<–70 °C (liquid)
  • 2…8 °C (lyophilised);<–70 °C (liquid)
  • 2…8 °C (lyophilised);<–70 °C (liquid)
  • Shipping temperature:
  • 25 °C
  • 25 °C
  • 25 °C
  • UniProtKB:
  • Uniprot # P07174
  • Uniprot # P07174
  • Uniprot # P07174

 

Frequently Bought Together