Cerium(IV) sulphate 0.05 mol/l (0.05 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # VWRS31307.460
Supplier: VWR Collection
CAS Number:
Specifications
- Pack type:Plastic container
- Pk:25 L
- Seq:5327
Specifications
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):0.0499 - 0.0501 mol/l
- NIST traceable:Confirmed
Specification Test Results
About this item
:
:332.24 g/mol
Merck Index:13,02002
EINECS:237-029-5
UN:3264
ADR:8,III
:GHS07
CAS Number:13590-82-4
Formula:Ce(SO₄)₂
MDL Number:MFCD00148852