

2-Propanol ≥99.0% Ph. Eur., USP
Catalog # VWRS20904.470
Supplier: VWR Collection
CAS Number:
Specifications
- Pk:25 L
- Seq:5327
Specifications
Specification Test Results
- Assay:Min. 99.0 %
- Appearance:Clear colourless liquid
- Relative density:0.785 - 0.789
- Specific gravity:0.783 - 0.787
- Refractive index (20°C):1.376 - 1.378
- Identification (IR):Passes test USP
- Identification B (*):Passes test USP
- Appearance test:Passes test Ph.Eur.
- Acidity or alkalinity:Passes test
- Absorbance (230 nm):Max. 0.3
- Absorbance (250 nm):Max. 0.1
- Absorbance (270 nm):Max. 0.03
- Absorbance (290 nm):Max. 0.02
- Absorbance (310 nm):Max. 0.01
- The absorption curve is smooth:Passes test Ph.Eur.
- Benzene:Max. 2 ppm
- Total of impurities apart from 2-butanol:Max. 0.3 %
- Peroxides:Passes test Ph.Eur.
- Non-volatile substances:Max. 20 ppm
- Water:Max. 0.5 %
- Each individual impurity (*):Max. 0.1 %
- Total impurities (*):Max. 1 %
- Residual solvents (CPMP/ICH/283/95):Conforms
- Residues of metal catalysts or reagents:Conforms
- Conforms to Ph.Eur.:Passes test
- Conforms to USP:Passes test
Specification Test Results
About this item
:
:60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0,786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
REACH:01-2119457558-25
:GHS02
:GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674