Order Entry
Ireland
ContactUsLinkComponent
Columns, HiTrap™ Macrocap SP
Columns, HiTrap™ Macrocap SP
Catalog # GEHE28-9508-59
Supplier:  Cytiva
Columns, HiTrap™ Macrocap SP
Catalog # GEHE28-9508-59
Supplier:  Cytiva

Specifications

  • Capacity:
    5 ml
  • Description:
    HiTrap™ MacroCap SP
  • Storage:
    4 to 30 °C, 20% Ethanol, 0,2 M Sodium Acetate
  • Bed dimensions:
    16×25 mm
  • Pk:
    5

Specifications

About this item