Order Entry
Ireland
ContactUsLinkComponent
VWR® PerfectBlue Twin ExW S, Dual-Gel System
VWR® PerfectBlue Twin ExW S, Dual-Gel System
Supplier:  VWR Collection
Certificates
undefined
VWR® PerfectBlue Twin ExW S, Dual-Gel System
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • 700-0986
  • 700-0980
  • Buffer volume, gel tank:
  • ~650 ml
  • ~650 ml
  • Sample capacity:
  • 10 − 72
  • 10 − 72
  • Unit W×H×D:
  • 300×130×140 mm
  • 300×130×140 mm
Specifications
Combs
Product Information
Frequently Bought Together