Paramat Extra, Solidification point: 55 - 58 °C Paramat extra, in pastille form, Gurr™, with DMSO
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:8002-74-2
:8012-95-1
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance (molten after 7 days; 65°C):Clear
- Cutting/ribboning:Passes test
- Congealing temperature:55 - 58 °C
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC361334C spec_VWRC361334C spec_VWRC361334C spec_VWRC361336E spec_VWRC361336E
Product specification
More
Frequently Bought Together