Order Entry
Ireland
Orders LinkContactUsLinkComponent
 

Human Recombinant IFNg (from E. coli)

Human Recombinant IFNg (from <i>E. coli</i>)
 
Human Recombinant IFNg (from <i>E. coli</i>)
Human Recombinant IFNg (from E. coli)

 

 

Interferon gamma (IFN-γ) is a type II interferon that is critical during adaptive and innate immune responses to infection. IFN-γ is produced by T cells and natural killer cells following antigen-specific activation. IFN-γ binds IFN-γ receptors (IFN-γ R1 and IFN-γ R2), which are expressed on most immune cells, to activate the JAK-STAT pathway. IFN-γ-induced signaling increases the expression of class 1 major histocompatibility complex (MHC) molecules. Human IFN-γ is not cross-reactive with mouse IFN-γ.

  • High Quality
  • Low Endotoxin
  • Affordable

A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  

 : For research use only.

 

 

 

  • Catalog No:
  • SHBT100-77-20UG
  • SHBT100-77-100UG
  • SHBT100-77-250UG
  • SHBT100-77-500UG
  • SHBT100-77-1MG
  • Protein/peptide name:
  • IFNg
  • IFNg
  • IFNg
  • IFNg
  • IFNg
  • Protein synonyms:
  • Type II interferon|T cell interferon|MAF|Immune Interferon
  • Type II interferon|T cell interferon|MAF|Immune Interferon
  • Type II interferon|T cell interferon|MAF|Immune Interferon
  • Type II interferon|T cell interferon|MAF|Immune Interferon
  • Type II interferon|T cell interferon|MAF|Immune Interferon
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Human
  • Human
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • P01579
  • P01579
  • P01579
  • P01579
  • P01579
  • Sequence:
  • MQDPYVKEAE NLKKYFNAGH SDVADNGTLF LGILKNWKEE SDRKIMQSQI VSFYFKLFKN FKDDQSIQKS VETIKEDMNV KFFNSNKKKR DDFEKLTNYS VTDLNVQRKA IHELIQVMAE LSPAAKTGKR KRSQMLFQGR RASQ
  • MQDPYVKEAE NLKKYFNAGH SDVADNGTLF LGILKNWKEE SDRKIMQSQI VSFYFKLFKN FKDDQSIQKS VETIKEDMNV KFFNSNKKKR DDFEKLTNYS VTDLNVQRKA IHELIQVMAE LSPAAKTGKR KRSQMLFQGR RASQ
  • MQDPYVKEAE NLKKYFNAGH SDVADNGTLF LGILKNWKEE SDRKIMQSQI VSFYFKLFKN FKDDQSIQKS VETIKEDMNV KFFNSNKKKR DDFEKLTNYS VTDLNVQRKA IHELIQVMAE LSPAAKTGKR KRSQMLFQGR RASQ
  • MQDPYVKEAE NLKKYFNAGH SDVADNGTLF LGILKNWKEE SDRKIMQSQI VSFYFKLFKN FKDDQSIQKS VETIKEDMNV KFFNSNKKKR DDFEKLTNYS VTDLNVQRKA IHELIQVMAE LSPAAKTGKR KRSQMLFQGR RASQ
  • MQDPYVKEAE NLKKYFNAGH SDVADNGTLF LGILKNWKEE SDRKIMQSQI VSFYFKLFKN FKDDQSIQKS VETIKEDMNV KFFNSNKKKR DDFEKLTNYS VTDLNVQRKA IHELIQVMAE LSPAAKTGKR KRSQMLFQGR RASQ
  • Protease-free:
  • N
  • N
  • N
  • N
  • N
  • Animal-free:
  • N
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
  • Bioactivity assay: Viral challenge assay using EMC virus on A549 cells, ED50 ≤0,75 ng/ml, Bioactivity ≥1,3×10⁶ units/mg
  • Bioactivity assay: Viral challenge assay using EMC virus on A549 cells, ED50 ≤0,75 ng/ml, Bioactivity ≥1,3×10⁶ units/mg
  • Bioactivity assay: Viral challenge assay using EMC virus on A549 cells, ED50 ≤0,75 ng/ml, Bioactivity ≥1,3×10⁶ units/mg
  • Bioactivity assay: Viral challenge assay using EMC virus on A549 cells, ED50 ≤0,75 ng/ml, Bioactivity ≥1,3×10⁶ units/mg
  • Bioactivity assay: Viral challenge assay using EMC virus on A549 cells, ED50 ≤0,75 ng/ml, Bioactivity ≥1,3×10⁶ units/mg
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
  • 10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
  • 10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
  • 10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
  • 10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
  • Molecular weight:
  • 16,9 kDa
  • 16,9 kDa
  • 16,9 kDa
  • 16,9 kDa
  • 16,9 kDa
  • Reconstitution instructions:
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Storage conditions:
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT
  • RT
  • Tested applications:
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay

 

Product Family Options

 

  • Pack type
    Pk
     
     
  • Vial
    20 µG
       
       
    1 * 20 µG
    1 * 20 µG - 76,50 €
    76,50 €
    1 * 20 µG
       76,50 €

     
     
    Specifications
    Cat. No.
    SHBT100-77-20UG
    Protein/peptide name
    IFNg
    Protein synonyms
    Type II interferon|T cell interferon|MAF|Immune Interferon
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P01579
    Sequence
    MQDPYVKEAE NLKKYFNAGH SDVADNGTLF LGILKNWKEE SDRKIMQSQI VSFYFKLFKN FKDDQSIQKS VETIKEDMNV KFFNSNKKKR DDFEKLTNYS VTDLNVQRKA IHELIQVMAE LSPAAKTGKR KRSQMLFQGR RASQ
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: Viral challenge assay using EMC virus on A549 cells, ED50 ≤0,75 ng/ml, Bioactivity ≥1,3×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
    Molecular weight
    16,9 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    100 µG
       
       
    1 * 100 µG
    1 * 100 µG - 190,00 €
    190,00 €
    1 * 100 µG
       190,00 €

     
     
    Specifications
    Cat. No.
    SHBT100-77-100UG
    Protein/peptide name
    IFNg
    Protein synonyms
    Type II interferon|T cell interferon|MAF|Immune Interferon
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P01579
    Sequence
    MQDPYVKEAE NLKKYFNAGH SDVADNGTLF LGILKNWKEE SDRKIMQSQI VSFYFKLFKN FKDDQSIQKS VETIKEDMNV KFFNSNKKKR DDFEKLTNYS VTDLNVQRKA IHELIQVMAE LSPAAKTGKR KRSQMLFQGR RASQ
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: Viral challenge assay using EMC virus on A549 cells, ED50 ≤0,75 ng/ml, Bioactivity ≥1,3×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
    Molecular weight
    16,9 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    250 µG
       
       
    1 * 250 µG
    1 * 250 µG - 437,00 €
    437,00 €
    1 * 250 µG
       437,00 €

     
     
    Specifications
    Cat. No.
    SHBT100-77-250UG
    Protein/peptide name
    IFNg
    Protein synonyms
    Type II interferon|T cell interferon|MAF|Immune Interferon
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P01579
    Sequence
    MQDPYVKEAE NLKKYFNAGH SDVADNGTLF LGILKNWKEE SDRKIMQSQI VSFYFKLFKN FKDDQSIQKS VETIKEDMNV KFFNSNKKKR DDFEKLTNYS VTDLNVQRKA IHELIQVMAE LSPAAKTGKR KRSQMLFQGR RASQ
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: Viral challenge assay using EMC virus on A549 cells, ED50 ≤0,75 ng/ml, Bioactivity ≥1,3×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
    Molecular weight
    16,9 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    500 µG
       
       
    1 * 500 µG
    1 * 500 µG - 729,00 €
    729,00 €
    1 * 500 µG
       729,00 €

     
     
    Specifications
    Cat. No.
    SHBT100-77-500UG
    Protein/peptide name
    IFNg
    Protein synonyms
    Type II interferon|T cell interferon|MAF|Immune Interferon
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P01579
    Sequence
    MQDPYVKEAE NLKKYFNAGH SDVADNGTLF LGILKNWKEE SDRKIMQSQI VSFYFKLFKN FKDDQSIQKS VETIKEDMNV KFFNSNKKKR DDFEKLTNYS VTDLNVQRKA IHELIQVMAE LSPAAKTGKR KRSQMLFQGR RASQ
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: Viral challenge assay using EMC virus on A549 cells, ED50 ≤0,75 ng/ml, Bioactivity ≥1,3×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
    Molecular weight
    16,9 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    1 mg
       
       
    1 * 1 mg
    1 * 1 mg - 1 270,00 €
    1 270,00 €
    1 * 1 mg
       1 270,00 €

     
     
    Specifications
    Cat. No.
    SHBT100-77-1MG
    Protein/peptide name
    IFNg
    Protein synonyms
    Type II interferon|T cell interferon|MAF|Immune Interferon
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P01579
    Sequence
    MQDPYVKEAE NLKKYFNAGH SDVADNGTLF LGILKNWKEE SDRKIMQSQI VSFYFKLFKN FKDDQSIQKS VETIKEDMNV KFFNSNKKKR DDFEKLTNYS VTDLNVQRKA IHELIQVMAE LSPAAKTGKR KRSQMLFQGR RASQ
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: Viral challenge assay using EMC virus on A549 cells, ED50 ≤0,75 ng/ml, Bioactivity ≥1,3×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
    Molecular weight
    16,9 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay