


HMDS (1,1,1,3,3,3-hexamethyldisilazane) 99+%, Electronic Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:999-97-3
:
Formula:(CH₃)₃SiNHSi(CH₃)₃
MDL Number:MFCD00008259
:161.39 g/mol
Merck Index:14,4689
Boiling Pt:126 °C
Melting Pt:–78 °C
Density:0,774
Flash Pt:8 °C (46 °F)
Storage Temperature:Ambient
EINECS:213-668-5
UN:3286
ADR:3,II
REACH:01-2119438176-38
:GHS02
:GHS06
:GHS05
Related Information
Frequently Bought Together