Order Entry
Ireland
Orders LinkContactUsLinkComponent
 

Human Recombinant GM-CSF (from E. coli)

Human Recombinant GM-CSF (from <i>E. coli</i>)
 
Human Recombinant GM-CSF (from <i>E. coli</i>)
Human Recombinant GM-CSF (from E. coli)

 

 

Granulocyte-macrophage colony-stimulating factor (GM-CSF) is hematopoietic growth factor produced by endothelial cells, monocytes, fibroblasts, and T cells. GM-CSF stimulates the production of neutrophilic granulocytes, macrophages, and mixed granulocyte-macrophage colonies from bone marrow cells. GM-CSF promotes immune system development and regulates neutrophil function during infection. Human and mouse GM-CSF show no cross-reactivity.

  • High Quality
  • Low Endotoxin
  • Affordable

A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  

 : For research use only.

 

 

 

 

  • Catalog No:
  • SHBT100-08-5UG
  • SHBT100-08-20UG
  • SHBT100-08-100UG
  • SHBT100-08-500UG
  • SHBT100-08-1MG
  • Protein/peptide name:
  • GM-CSF
  • GM-CSF
  • GM-CSF
  • GM-CSF
  • GM-CSF
  • Protein synonyms:
  • MGI1GM|CSF-2|Pluripoietin-α
  • MGI1GM|CSF-2|Pluripoietin-α
  • MGI1GM|CSF-2|Pluripoietin-α
  • MGI1GM|CSF-2|Pluripoietin-α
  • MGI1GM|CSF-2|Pluripoietin-α
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Human
  • Human
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • P04141
  • P04141
  • P04141
  • P04141
  • P04141
  • Sequence:
  • MAPARSPSPS TQPWEHVNAI QEARRLLNLS RDTAAEMNET VEVISEMFDL QEPTCLQTRL ELYKQGLRGS LTKLKGPLTM MASHYKQHCP PTPETSCATQ IITFESFKEN LKDFLLVIPF DCWEPVQE
  • MAPARSPSPS TQPWEHVNAI QEARRLLNLS RDTAAEMNET VEVISEMFDL QEPTCLQTRL ELYKQGLRGS LTKLKGPLTM MASHYKQHCP PTPETSCATQ IITFESFKEN LKDFLLVIPF DCWEPVQE
  • MAPARSPSPS TQPWEHVNAI QEARRLLNLS RDTAAEMNET VEVISEMFDL QEPTCLQTRL ELYKQGLRGS LTKLKGPLTM MASHYKQHCP PTPETSCATQ IITFESFKEN LKDFLLVIPF DCWEPVQE
  • MAPARSPSPS TQPWEHVNAI QEARRLLNLS RDTAAEMNET VEVISEMFDL QEPTCLQTRL ELYKQGLRGS LTKLKGPLTM MASHYKQHCP PTPETSCATQ IITFESFKEN LKDFLLVIPF DCWEPVQE
  • MAPARSPSPS TQPWEHVNAI QEARRLLNLS RDTAAEMNET VEVISEMFDL QEPTCLQTRL ELYKQGLRGS LTKLKGPLTM MASHYKQHCP PTPETSCATQ IITFESFKEN LKDFLLVIPF DCWEPVQE
  • Protease-free:
  • N
  • N
  • N
  • N
  • N
  • Animal-free:
  • N
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
  • Bioactivity assay: TF-1 cell proliferation, ED50 ≤0,2 ng/ml, Bioactivity ≥5,0×10⁶ units/mg
  • Bioactivity assay: TF-1 cell proliferation, ED50 ≤0,2 ng/ml, Bioactivity ≥5,0×10⁶ units/mg
  • Bioactivity assay: TF-1 cell proliferation, ED50 ≤0,2 ng/ml, Bioactivity ≥5,0×10⁶ units/mg
  • Bioactivity assay: TF-1 cell proliferation, ED50 ≤0,2 ng/ml, Bioactivity ≥5,0×10⁶ units/mg
  • Bioactivity assay: TF-1 cell proliferation, ED50 ≤0,2 ng/ml, Bioactivity ≥5,0×10⁶ units/mg
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 10 mM sodium phosphate, pH 7,5
  • 10 mM sodium phosphate, pH 7,5
  • 10 mM sodium phosphate, pH 7,5
  • 10 mM sodium phosphate, pH 7,5
  • 10 mM sodium phosphate, pH 7,5
  • Molecular weight:
  • 14,6 kDa
  • 14,6 kDa
  • 14,6 kDa
  • 14,6 kDa
  • 14,6 kDa
  • Reconstitution instructions:
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Storage conditions:
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT
  • RT
  • Tested applications:
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay

 

Product Family Options

 

  • Pack type
    Pk
     
     
  • Vial
    5 µG
       
       
    1 * 5 µG
    1 * 5 µG - 76,50 €
    76,50 €
    1 * 5 µG
       76,50 €

     
     
    Specifications
    Cat. No.
    SHBT100-08-5UG
    Protein/peptide name
    GM-CSF
    Protein synonyms
    MGI1GM|CSF-2|Pluripoietin-α
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P04141
    Sequence
    MAPARSPSPS TQPWEHVNAI QEARRLLNLS RDTAAEMNET VEVISEMFDL QEPTCLQTRL ELYKQGLRGS LTKLKGPLTM MASHYKQHCP PTPETSCATQ IITFESFKEN LKDFLLVIPF DCWEPVQE
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: TF-1 cell proliferation, ED50 ≤0,2 ng/ml, Bioactivity ≥5,0×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, pH 7,5
    Molecular weight
    14,6 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    20 µG
       
       
    1 * 20 µG
    1 * 20 µG - 190,00 €
    190,00 €
    1 * 20 µG
       190,00 €

     
     
    Specifications
    Cat. No.
    SHBT100-08-20UG
    Protein/peptide name
    GM-CSF
    Protein synonyms
    MGI1GM|CSF-2|Pluripoietin-α
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P04141
    Sequence
    MAPARSPSPS TQPWEHVNAI QEARRLLNLS RDTAAEMNET VEVISEMFDL QEPTCLQTRL ELYKQGLRGS LTKLKGPLTM MASHYKQHCP PTPETSCATQ IITFESFKEN LKDFLLVIPF DCWEPVQE
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: TF-1 cell proliferation, ED50 ≤0,2 ng/ml, Bioactivity ≥5,0×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, pH 7,5
    Molecular weight
    14,6 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    100 µG
       
       
    1 * 100 µG
    1 * 100 µG - 476,00 €
    476,00 €
    1 * 100 µG
       476,00 €

     
     
    Specifications
    Cat. No.
    SHBT100-08-100UG
    Protein/peptide name
    GM-CSF
    Protein synonyms
    MGI1GM|CSF-2|Pluripoietin-α
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P04141
    Sequence
    MAPARSPSPS TQPWEHVNAI QEARRLLNLS RDTAAEMNET VEVISEMFDL QEPTCLQTRL ELYKQGLRGS LTKLKGPLTM MASHYKQHCP PTPETSCATQ IITFESFKEN LKDFLLVIPF DCWEPVQE
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: TF-1 cell proliferation, ED50 ≤0,2 ng/ml, Bioactivity ≥5,0×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, pH 7,5
    Molecular weight
    14,6 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    500 µG
       
       
    1 * 500 µG
    1 * 500 µG - 2 130,00 €
    2 130,00 €
    1 * 500 µG
       2 130,00 €

     
     
    Specifications
    Cat. No.
    SHBT100-08-500UG
    Protein/peptide name
    GM-CSF
    Protein synonyms
    MGI1GM|CSF-2|Pluripoietin-α
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P04141
    Sequence
    MAPARSPSPS TQPWEHVNAI QEARRLLNLS RDTAAEMNET VEVISEMFDL QEPTCLQTRL ELYKQGLRGS LTKLKGPLTM MASHYKQHCP PTPETSCATQ IITFESFKEN LKDFLLVIPF DCWEPVQE
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: TF-1 cell proliferation, ED50 ≤0,2 ng/ml, Bioactivity ≥5,0×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, pH 7,5
    Molecular weight
    14,6 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    1 mg
       
       
    1 * 1 mg
    1 * 1 mg - 3 550,00 €
    3 550,00 €
    1 * 1 mg
       3 550,00 €

     
     
    Specifications
    Cat. No.
    SHBT100-08-1MG
    Protein/peptide name
    GM-CSF
    Protein synonyms
    MGI1GM|CSF-2|Pluripoietin-α
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P04141
    Sequence
    MAPARSPSPS TQPWEHVNAI QEARRLLNLS RDTAAEMNET VEVISEMFDL QEPTCLQTRL ELYKQGLRGS LTKLKGPLTM MASHYKQHCP PTPETSCATQ IITFESFKEN LKDFLLVIPF DCWEPVQE
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: TF-1 cell proliferation, ED50 ≤0,2 ng/ml, Bioactivity ≥5,0×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, pH 7,5
    Molecular weight
    14,6 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.