Order Entry
Ireland
Orders LinkContactUsLinkComponent
Mouse Recombinant RELM-A (from <i>E. coli</i>)

Mouse Recombinant RELM-A (from E. coli)

Mouse Recombinant RELM-A (from <i>E. coli</i>)
Certificates
Mouse Recombinant RELM-A (from <i>E. coli</i>)

5 Options Available Below

Product Details & Documents

Thrombopoietin (TPO) is a growth factor that is produced by liver and kidney tissues. TPO binds the TPO receptor (CD110) to promote megakaryocyte maturation, differentiation, and the production of platelets.

  • High quality
  • Low endotoxin
  • Affordable

Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.

Delivery information: Certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.

Caution: For research use only.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SHBT200-28-5UG
  • SHBT200-28-25UG
  • SHBT200-28-100UG
  • SHBT200-28-500UG
  • SHBT200-28-1MG
  • Protein/peptide name:
  • RELM-alpha
  • RELM-alpha
  • RELM-alpha
  • RELM-alpha
  • RELM-alpha
  • Protein synonyms:
  • FIZZ1
  • FIZZ1
  • FIZZ1
  • FIZZ1
  • FIZZ1
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • Q9EP95
  • Q9EP95
  • Q9EP95
  • Q9EP95
  • Q9EP95
  • Sequence:
  • MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
  • MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
  • MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
  • MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
  • MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
  • Protease-free:
  • N
  • N
  • N
  • N
  • N
  • Animal-free:
  • N
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤1
  • Endotoxin LAL (EU/µg) ≤1
  • Endotoxin LAL (EU/µg) ≤1
  • Endotoxin LAL (EU/µg) ≤1
  • Endotoxin LAL (EU/µg) ≤1
  • UniProtKB:
  • Q9EP95
  • Q9EP95
  • Q9EP95
  • Q9EP95
  • Q9EP95
  • Biological activity:
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 10 mM sodium phosphate, pH 7,5
  • 10 mM sodium phosphate, pH 7,5
  • 10 mM sodium phosphate, pH 7,5
  • 10 mM sodium phosphate, pH 7,5
  • 10 mM sodium phosphate, pH 7,5
  • Molecular weight:
  • 9,6 kDa
  • 9,6 kDa
  • 9,6 kDa
  • 9,6 kDa
  • 9,6 kDa
  • Reconstitution instructions:
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Storage conditions:
  • −20 °C
  • −20 °C
  • −20 °C
  • −20 °C
  • −20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT
  • RT

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Vial
    5 µG
    Log in for availability
    Log in for availability
    1 * 5 µG
    1 * 5 µG - 76,50 €
    76,50 €
    1 * 5 µG
       76,50 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-28-5UG
    Protein/peptide name
    RELM-alpha
    Protein synonyms
    FIZZ1
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q9EP95
    Sequence
    MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤1
    UniProtKB
    Q9EP95
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, pH 7,5
    Molecular weight
    9,6 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • Vial
    25 µG
    Log in for availability
    Log in for availability
    1 * 25 µG
    1 * 25 µG - 190,00 €
    190,00 €
    1 * 25 µG
       190,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-28-25UG
    Protein/peptide name
    RELM-alpha
    Protein synonyms
    FIZZ1
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q9EP95
    Sequence
    MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤1
    UniProtKB
    Q9EP95
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, pH 7,5
    Molecular weight
    9,6 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • Vial
    100 µG
    Log in for availability
    Log in for availability
    1 * 100 µG
    1 * 100 µG - 476,00 €
    476,00 €
    1 * 100 µG
       476,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-28-100UG
    Protein/peptide name
    RELM-alpha
    Protein synonyms
    FIZZ1
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q9EP95
    Sequence
    MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤1
    UniProtKB
    Q9EP95
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, pH 7,5
    Molecular weight
    9,6 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • Vial
    500 µG
    Log in for availability
    Log in for availability
    1 * 500 µG
    1 * 500 µG - 2 060,00 €
    2 060,00 €
    1 * 500 µG
       2 060,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-28-500UG
    Protein/peptide name
    RELM-alpha
    Protein synonyms
    FIZZ1
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q9EP95
    Sequence
    MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤1
    UniProtKB
    Q9EP95
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, pH 7,5
    Molecular weight
    9,6 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • Vial
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 3 420,00 €
    3 420,00 €
    1 * 1 mg
       3 420,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-28-1MG
    Protein/peptide name
    RELM-alpha
    Protein synonyms
    FIZZ1
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q9EP95
    Sequence
    MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤1
    UniProtKB
    Q9EP95
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, pH 7,5
    Molecular weight
    9,6 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT