Pyrene for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:129-00-0
:
Formula:C₁₆H₁₀
MDL Number:MFCD00004136
:202.26 g/mol
Merck Index:12,08147
Boiling Pt:404 °C (1013 hPa)
Melting Pt:148…150 °C
Density:1,27 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-927-3
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ200
Flash Pt:200…220 °C
:GHS09
Specification Test Results
- Solubility:0.135 mg/l (25 °C)
- Melting Point:148 - 150 °C
- Molar Mass:202.26 g/mol
- Bulk Density:650 kg/m3
- Boiling Point:393 °C (1013 hPa)
- Flash Point:200 - 220 °C
- Density:1.21 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together