Order Entry
Ireland
Orders LinkContactUsLinkComponent
 

Adenine

Toxic
Marketsource
Supplier:   Roth Carl
Adenine
Supplier:  Roth Carl
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine
  • CAS Number:73-24-5
  • Formula:C₅H₅N₅
  • MDL Number:MFCD00041790
  • Molecular Weight:135.13 g/mol
  • Merck Index:13,00152
  • Density:0,99741 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:200-796-1
  • UN:2811
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.