Order Entry
Ireland
ContactUsLinkComponent
Succinic anhydride for synthesis, Sigma-Aldrich®
Corrosive
Health
Irritant
Succinic anhydride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Health
Irritant
Succinic anhydride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    67 g/l (20 °C) Hydrolysis
  • Melting Point:
    116 - 120 °C
  • Molar Mass:
    100.07 g/mol
  • Bulk Density:
    550 - 600 kg/m3
  • Boiling Point:
    261 °C (1013 hPa)
  • Vapor Pressure:
    1.3 hPa (92 °C)
  • Flash Point:
    157 °C
  • Density:
    1.48 g/cm3 (20 °C)
  • pH:
    2.7 (20 g/l, H2O)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together