Order Entry
Ireland
ContactUsLinkComponent
 
Irritant
Iron(II,III) oxide, black, TECHNICAL
Catalog # 24187.460
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Iron(II,III) oxide, black, TECHNICAL
Catalog # 24187.460
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Metal drum
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Identification:
    Passes test

Specification Test Results

About this item

 :
 :231.54 g/mol
Merck Index:12,04087
Melting Pt:1560 °C
Density:5,18 g/cm³ (20 °C)
EINECS:215-277-5
 :GHS07
CAS Number:1317-61-9
Formula:Fe₃O₄
MDL Number:MFCD00011010