

Chromium(II) chloride, anhydrous 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10049-05-5
:
Formula:Cl₂Cr
MDL Number:MFCD00010947
:122.9 g/mol
Merck Index:14,2244
Boiling Pt:1302 °C (1013 hPa)
Melting Pt:824 °C
Density:2,878
Storage Temperature:Ambient
EINECS:233-163-3
UN:3260
ADR:8,III
:GHS05
:GHS07
Related Information
Frequently Bought Together