Tantalum pentachloride, anhydrous ≥99.995% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7721-01-9
:
Formula:Cl₅Ta
MDL Number:MFCD00011253
:358.21 g/mol
Merck Index:14,9054
Boiling Pt:242 °C
Melting Pt:216 °C
Density:3,68
UN:3260
ADR:8,III
:ampouled under argon
EINECS:231-755-6
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together