Order Entry
Ireland
ContactUsLinkComponent
Titanium(IV) isopropoxide
Titanium(IV) isopropoxide
Catalog # MOLE33520313-500G
Supplier:  Molekula
CAS Number:  
Titanium(IV) isopropoxide
Catalog # MOLE33520313-500G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Titanium tetraisopropanolate, Titanium (IV) isopropoxide
 :
 :284.23 g/mol
EINECS:208-909-6
UN:2413
CAS Number:546-68-9
Formula:C₁₂H₂₈O₄Ti
MDL Number:MFCD00008871
Boiling Pt:240
Melting Pt:18…20 °C
Density:0.9711 g/cm³ (20 °C)
Flash Pt:51
ADR:3 III
Merck Index:12,09614