Titanium(IV) chloride 99.9%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7550-45-0
:
Formula:Cl₄Ti
MDL Number:MFCD00011267
:189.69 g/mol
Merck Index:13,09555
Boiling Pt:136 °C (1013 hPa)
Melting Pt:–24 °C
Density:1,72 g/cm³
Storage Temperature:Ambient
EINECS:231-441-9
UN:1838
ADR:6.1,I
:GHS05
:GHS06
Specification Test Results
- Appearance:Clear light yellow Liquid
- Assay:≥99.85 %
- Non-volatile matter:≤500 ppm
- Aluminium (Al):≤2 ppm
- Arsenic (As):≤2 ppm
- Carbon (C):≤50 ppm
- Iron (Fe):≤1 ppm
- Silicon (Si):≤50 ppm
- Vanadium (V):≤5 ppm
Specification Test Results
Frequently Bought Together