Diyttrium trioxide ≥99.9% (REO, rare earth oxide basis), pellets
Catalog # 40648.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
CAS Number:1314-36-9
:
Formula:Y₂O₃
MDL Number:MFCD00011473
Merck Index:14,10107
Boiling Pt:4300 °C (1013 hPa)
Melting Pt:2410 °C
Density:5,01
:225.81 g/mol
Storage Temperature:Ambient
EINECS:215-233-5