Mercury(II) iodide, red ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
:7774-29-0
:
:HgI₂
:MFCD00011044
:454.4 g/mol
:13,05905
:354 °C (1013 hPa)
:259 °C
:6,36 g/cm³ (20 °C)
:Ambient
:1638
:6.1,II
:M-Clarity™ quality level = MQ300
:231-873-8
:GHS06
:GHS08
:GHS09
- Assay (argentometric, dried sust.):≥ 99.0 %
- Solubility in Potassium iodide:passes test
- Soluble mercury salts (as Hg):≤ 0.05 %
- Foreign heavy metals (as Pb):≤ 0.001 %
- Fe (Iron):≤ 0.001 %
- Mercury (I) (as Hg):≤ 0.1 %
- Substances not reducing by formic acid:≤ 0.02 %
- Corresponds to ACS,Reag. Ph Eur:6 - 7 (50 g/l, H2O, 20 °C) (slurry)
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Frequently Bought Together