Mercury(II) iodide ≥99.0% (dried basis) ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7774-29-0
:
Formula:HgI₂
MDL Number:MFCD00011044
:454.4 g/mol
Merck Index:14,5879
Boiling Pt:350 °C (subl.)
Melting Pt:259 °C
Density:6,28
Storage Temperature:Ambient
EINECS:231-873-8
UN:1638
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Frequently Bought Together