Order Entry
Ireland
ContactUsLinkComponent
Mercury(II) iodide, red 99+%
Mercury(II) iodide, red 99+%
Certificates
undefined
Mercury(II) iodide, red 99+%
Certificates

Specification Test Results

  • Appearance:
    Red Crystalline powder
  • Titration Iodometric:
    ≥99.0 %
  • Solubility:
    (in Potassium iodide soln.) Passes test
  • Mercurous Mercury (as Hg):
    ≤0.1 %
  • Soluble Hg salts (as Hg):
    ≤0.05 %

Specification Test Results

Related Information

Frequently Bought Together