


Zinc chloride
Supplier: Roth Carl
Certificates
About this item
CAS Number:7646-85-7
:
Formula:ZnCl₂
MDL Number:MFCD00011295
:136.3 g/mol
Merck Index:13,10185
Boiling Pt:732 °C (1013 hPa)
Melting Pt:283 °C
Density:2,91 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-592-0
UN:2331
ADR:8,III
:GHS05
:GHS07
:GHS09
Frequently Bought Together