Silver sulphate ≥98.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Disilver sulphate
CAS Number:10294-26-5
:
Formula:Ag₂SO₄
MDL Number:MFCD00003407
Boiling Pt:1085
Melting Pt:655
Density:5.45 g/cm³ (20 °C)
Molecular Weight:311.8 g/mol
EINECS:233-653-7
ADR:9 III
Merck Index:13,08603
UN:3077
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 98.5 %
- Solution in water:Passes test
- Foreign heavy metals (as Pb):Max. 20 ppm
- Insolubility in water:Max. 0.02 %
- Substances not precipitated by HCl:Max. 0.03 %
- Cl (Chloride):Max. 20 ppm
- Cu (Copper):Max. 5 ppm
- Fe (Iron):Max. 10 ppm
- Pb (Lead):Max. 10 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number250 ml Plastic bottle for solids DIN 60 (ZPACK13250250)
Packaging Information
Frequently Bought Together