Copper(I) iodide ≥98%
About this item
Purity: 98% CAS No.: 7681-65-4 Molecular Formula: CuI Formula Weight: 190.45 Physical Form: Powder Density: 5.63 Refractive Index: 2.346 Sensitivity: Light, Air and Moisture Sensitive Appearance: Pale brown Odour: Weak Melting Point: 605° Boiling Point: ca 1290° Solubility: Insoluble in water, dilute acids, alcohol. Decomposes by conc. acids. Soluble in NH3 solution, iodides, thiosulfates Merck Reference: 14,2662 UN No.: 3077 MDL No.: MFCD00010978
As a catalyst in organic synthesis, cathode ray tube coating, temperature indicator with HgI2
- CAS Number:7681-65-4
- Formula:CuI
- MDL Number:MFCD00010978
- Molecular Weight:190.45 g/mol
- Merck Index:14,2662
- Boiling Pt:∼1290 °C
- Melting Pt:605 °C
- Density:5,63
- Storage Temperature:Ambient
- EINECS:231-674-6
- UN:3077
- ADR:9,III
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 7681-65-4 Molecular Formula: CuI Formula Weight: 190.45 Physical Form: Powder Density: 5.63 Refractive Index: 2.346 Sensitivity: Light, Air and Moisture Sensitive Appearance: Pale brown Odour: Weak Melting Point: 605° Boiling Point: ca 1290° Solubility: Insoluble in water, dilute acids, alcohol. Decomposes by conc. acids. Soluble in NH3 solution, iodides, thiosulfates Merck Reference: 14,2662 UN No.: 3077 MDL No.: MFCD00010978
As a catalyst in organic synthesis, cathode ray tube coating, temperature indicator with HgI2
- CAS Number:7681-65-4
- Formula:CuI
- MDL Number:MFCD00010978
- Molecular Weight:190.45 g/mol
- Merck Index:14,2662
- Boiling Pt:∼1290 °C
- Melting Pt:605 °C
- Density:5,63
- Storage Temperature:Ambient
- EINECS:231-674-6
- UN:3077
- ADR:9,III



