Order Entry
Ireland
Orders LinkContactUsLinkComponent
 

4-Methylindole 99%

Irritant
4-Methylindole 99%
Certificates
4-Methylindole 99%

2 Options Available Below

Product Details & Documents
Synonyms:  4-Methyl-1H-indole
  • CAS Number:16096-32-5
  • Formula:C₉H₉N
  • MDL Number:MFCD00005668
  • Molecular Weight:131.18 g/mol
  • Boiling Pt:267 °C (760 mmHg)
  • Melting Pt:5 °C
  • Density:1,06 g/cm³
  • Flash Pt:>112 °C
  • Storage Temperature:Freezer
  • EINECS:240-262-5
  • UN:3334
  • ADR:9,III

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear yellow to brown Liquid after melting
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.6060 to 1.6080 (20°C, 589 nm)
  • GC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 273,00 €
    273,00 €
    1 * 1 g
       273,00 €

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 767,00 €
    767,00 €
    1 * 5 g
       767,00 €