Ammonium dihydrogen phosphate 99.9% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate
CAS Number:7722-76-1
:
Formula:NH₄H₂PO₄
MDL Number:MFCD00003396
Merck Index:13,00547
Melting Pt:190 °C
Density:1,8 g/cm³ (20 °C)
:115.03 g/mol
Storage Temperature:Ambient
EINECS:231-764-5
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Total trace metal impurities:≤0.1 %
Specification Test Results
Frequently Bought Together