Order Entry
Ireland
ContactUsLinkComponent
Ammonium fluoride ≥97%, GPR RECTAPUR®
 
Ammonium fluoride ≥97%, GPR RECTAPUR®
  21251.260
 :  
undefined
 
Ammonium fluoride ≥97%, GPR RECTAPUR®
  21251.260
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Assay:
    Min. 97 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Ignition residue (SO4):
    Max. 0.05 %
  • Fe (Iron):
    Max. 20 ppm

 

 

 :
 :37.04 g/mol
 :13,00524
 :160 °C
 :1,0088…1.0096 g/cm³ (25 °C)
 :Ambient
 :235-185-9
 :2505
 :6.1,III
 :GHS06
 :12125-01-8
 :NH₄F
 :MFCD00011423