Order Entry
Ireland
ContactUsLinkComponent
Tetrabutylammonium iodide for synthesis, Sigma-Aldrich®
Tetrabutylammonium iodide for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Tetrabutylammonium iodide for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Hill Formula:
    C₁₆H₃₆IN
  • Molar Mass:
    369.38 g/mol
  • Melting Point:
    143 - 146 °C
  • LD 50 oral:
    LD50 Rat 1990 mg/kg
  • Storage class:
    10 - 13 Other liquids and solids
  • WGK:
    WGK 3 highly water endangering
  • Categories of danger:
    harmful
  • R Phrase:
    R 22
  • S Phrase:
    S 22-24/25

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together