
(1-Pentyl)triphenylphosphonium bromide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:21406-61-1
:
Formula:H₃C(CH₂)₄P(C₆H₅)₃Br
MDL Number:MFCD00031630
:413.34 g/mol
Melting Pt:164…166 °C
EINECS:244-374-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together