Order Entry
Ireland
ContactUsLinkComponent
Triphenyl phosphate analytical standard, Supelco®
Environment
Triphenyl phosphate analytical standard, Supelco®
Catalog # 442829.
Supplier:  Merck
CAS Number:  
undefined
Environment
Triphenyl phosphate analytical standard, Supelco®
Catalog # 442829.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :326.29 g/mol
Merck Index:12,09872
Boiling Pt:412 °C (1013 hPa)
Melting Pt:50…51 °C
Density:1,33 g/cm³ (20 °C)
Flash Pt:223 °C
Storage Temperature:Ambient
EINECS:204-112-2
UN:3077
ADR:9,III
 :GHS09
CAS Number:115-86-6
Formula:C₁₈H₁₅O₄P
MDL Number:MFCD00003031