Order Entry
Ireland
ContactUsLinkComponent
 
 
 
Tributyl phosphate ≥99.0%, GPR RECTAPUR®
 
undefined
 
 
Tributyl phosphate ≥99.0%, GPR RECTAPUR®
 

 

  • Assay:
    Min. 99,0 %
  • Appearance:
    Clear liquid
  • Colouration:
    Max. 50 APHA
  • Density (20/4):
    0,975 --> 0,980
  • Refractive index (20°C):
    1,423 --> 1,425
  • Water:
    Max. 0,2 %

 

Frequently Bought Together