
Antimony(III) oxide ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1309-64-4
:
Formula:O₃Sb₂
MDL Number:MFCD00011214
:291.52 g/mol
Merck Index:14,711
Boiling Pt:1550 °C (subl.)
Melting Pt:656 °C
Density:5,67
Storage Temperature:Ambient
EINECS:215-175-0
UN:1549
ADR:6.1,III
:GHS08
Frequently Bought Together