Phenylsilane 97+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Silylbenzene
CAS Number:694-53-1
:
Formula:C₆H₈Si
MDL Number:MFCD00013478
Boiling Pt:120 °C (760 mmHg)
Density:0.87 g/cm³
Molecular Weight:108.22 g/mol
Flash Pt:7 °C
Storage Temperature:Ambient
EINECS:211-772-5
UN:3399
ADR:4.3 II
:GHS07
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5090 to 1.5120 (20°C, 589 nm)
- GC:≥97.0 %
Specification Test Results
Frequently Bought Together