

Tetraethyl orthosilicate (TEOS) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78-10-4
:
Formula:Si(OC₂H₅)₄
MDL Number:MFCD00009062
:208.33 g/mol
Merck Index:14,3851
Boiling Pt:168 °C
Melting Pt:–85 °C
Density:0,934
Flash Pt:46 °C (116 °F)
Storage Temperature:Ambient
EINECS:201-083-8
UN:1292
ADR:3,III
:GHS02
:GHS07
Related Information
Frequently Bought Together