Tetraethyl orthosilicate (TEOS) ≥99%
: Tetraethoxysilane
:78-10-4
:
:Si(OC₂H₅)₄
:MFCD00009062
:208.33 g/mol
:14,3851
:168 °C
:–85 °C
:0,934
:46 °C (116 °F)
:Ambient
:201-083-8
:1292
:3,III
:GHS02
:GHS07
- Environmentally Preferable:
Frequently Bought Together