Order Entry
Ireland
ContactUsLinkComponent
Aluminium sulphate octadecahydrate, AnalaR NORMAPUR® analytical reagent
Aluminium sulphate octadecahydrate, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Aluminium sulphate octadecahydrate, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (calculated as Al2(SO4)3):
    51.0 - 59.0 %
  • Solution (20 %; water):
    Clear colourless liquid
  • pH (20°C; 2 %):
    2.5 - 4.0
  • Alkali and alkaline-earth metals:
    Max. 0.4 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Cl (Chloride):
    Max. 50 ppm
  • NH4 (Ammonium):
    Max. 100 ppm
  • As (Arsenic):
    Max. 3 ppm
  • Fe (Iron):
    Max. 50 ppm

Specification Test Results

Related Information

Frequently Bought Together