Aluminium oxide 99.7+%, extra pure
:1344-28-1
:11092-32-3
:Al₂O₃
:MFCD00003424
:101.96 g/mol
:13,00355
:2977 °C (760 mmHg)
:2030 °C
:3,97 g/cm³
:Ambient
:215-691-6
:01-2119529248-35
- Appearance:White Powder
- Sodium oxide (Na2O):≤0.1 %
- Silicon dioxide (SiO2):≤0.04 %
- Aluminium oxide (Al2O3):≥99.70 %
- Iron(III)oxide (Fe2O3):≤0.03 %
Frequently Bought Together