Order Entry
Ireland
ContactUsLinkComponent
 
Magnesium sulphate, anhydrous ≥97%, Reagent Grade
Catalog # 208094-2.5KG
CAS Number:  
undefined
Magnesium sulphate, anhydrous ≥97%, Reagent Grade
Catalog # 208094-2.5KG
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

Specification Test Results

  • Assay of Mg:
    19.6 - 20.8 %
  • Vapor Pressure:
    <0.1 mmHg ( 20 °C)
  • Vapor Density:
    <0.01 (vs air)

Specification Test Results

About this item

 :
 :120.37 g/mol
Merck Index:12,05731
Melting Pt:1,130 °C (dec.)
Density:2,7 g/cm³
Storage Temperature:Ambient
EINECS:231-298-2
CAS Number:7487-88-9
Formula:MgSO₄
MDL Number:MFCD00011110