Order Entry
Ireland
ContactUsLinkComponent
Magnesium sulphate
Magnesium sulphate
Catalog # ROTH8283.1
Supplier:  Roth Carl
CAS Number:  
undefined
Magnesium sulphate
Catalog # ROTH8283.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

About this item

 :
 :120.37 g/mol
Merck Index:12,05731
Melting Pt:1127 °C
Density:2,65 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-298-2
CAS Number:7487-88-9
Formula:MgSO₄
MDL Number:MFCD00011110