Order Entry
Ireland
ContactUsLinkComponent
 
Magnesium hydroxide 95.0-100.5%, white powder USP, FCC, Macron Fine Chemicals™
 
undefined
Magnesium hydroxide 95.0-100.5%, white powder USP, FCC, Macron Fine Chemicals™
 

 

  • Environmentally Preferable:

 

 

  • Bulk Pharmaceutical Chemical:
    95.0 - 100.5 %
  • USP - Assay (Mg(OH)₂) (dried basis):
    98.0 - 102.0 %
  • USP - Calcium (Ca):
    ≤ 1.5 %
  • USP - Carbonate (CO₃):
    Passes Test
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • Lead (Pb)(USP):
    ≤ 1.5 ppm
  • USP - Loss on Drying at 105°C:
    ≤ 2.0 %
  • USP - Loss on Ignition:
    30.0 - 33.0 %
  • USP - Soluble Salts:
    ≤ 2.0 %
  • USP - Microbiological E Coli (negative):
    Passes Test
  • FCC - Assay (Mg(OH)₂) (dried basis):
    95.0 - 100.5 %
  • FCC - Identification:
    Passes Test
  • FCC - Loss on Drying at 105°C:
    ≤ 2.0 %
  • FCC - Loss on Ignition:
    30.0 - 33.0 %
  • FCC - Lead (Pb):
    ≤ 2 mg/kg
  • FCC - Calcium Oxide:
    ≤ 1 %
  • FCC - Alkalies (as SO₄):
    Passes Test

 

Frequently Bought Together