Calcium oxide ≥99.95% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1305-78-8
:
Formula:CaO
MDL Number:MFCD00010911
:56.08 g/mol
Merck Index:14,1686
Boiling Pt:2850 °C
Melting Pt:2572 °C
Density:3,37
Storage Temperature:Ambient
EINECS:215-138-9
UN:1910
ADR:8,III
:GHS05
:GHS07
Related Information
Frequently Bought Together