
Calcium fluoride 97%, Reagent Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7789-75-5
:
Formula:CaF₂
MDL Number:MFCD00010907
:78.07 g/mol
Merck Index:14,1667
Boiling Pt:∼2500 °C
Melting Pt:1403 °C
Density:3,18
EINECS:232-188-7
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together