Order Entry
Ireland
ContactUsLinkComponent
Sodium sulphate, anhydrous ≥99.0%, powder ACS
Sodium sulphate, anhydrous ≥99.0%, powder ACS
Supplier:  MP Biomedicals
Certificates
undefined
Sodium sulphate, anhydrous ≥99.0%, powder ACS
Supplier:  MP Biomedicals

Specification Test Results

  • Synonyms:
    Sodium sulphate (2:1); Disodium sulphate; Natriumsufat; Salt Cake; sulphuric acid, disodium salt; Thenardite; Trona
  • Form:
    White powder
  • Storage:
    Store at room temperature (15 to 30 °C)
  • Assay:
    99,0%
  • Heavy Metals:
    5 ppm (as Pb)
  • Insoluble Matter:
    0,01%
  • Loss on Drying:
    0,5%
  • pH:
    5,2 - 9,2 (5% aq soln)
  • Refractive Index:
    1,484 (Lit.)

Specification Test Results

Frequently Bought Together